Assay Design



Gene Transcripts

Gene Symbol
RAB8
Entrez Gene ID
824529
Gene Name
RAB GTPase homolog 8
Gene Aliases
AT3G53610, ATRAB8, AtRABE1a, AtRab8B, GTPASE ATRAB8, RAB GTPase homolog 8
Chromosome Location
Chr.3: 19876232 - 19878792 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084809.2 NP_001078278.1 5 - 6 942 67
NM_115221.4 NP_190929.1 6 - 7 819 67
NM_180365.3 NP_850696.1 6 - 7 492 67
GenBank mRNA AK228083.1 - 6 - 7 519 67
AK230120.1 - - 930 67
AY086756.1 - 6 - 7 529 67
BT004779.1 - 5 - 6 414 67
U82434.1 - 6 - 7 434 67

View Full Product Details