Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_120582.5 | NP_196119.1 | 2 - 3 | 274 | 76 |
NM_180437.2 | NP_850768.1 | 2 - 3 | 156 | 76 | |
NM_203003.3 | NP_974732.1 | 1 - 2 | 614 | 76 | |
GenBank mRNA | AK229425.1 | - | 2 - 3 | 217 | 76 |
BT044057.1 | - | 1 - 2 | 105 | 76 |