Assay Design



Gene Transcripts

Gene Symbol
KEG
Entrez Gene ID
831197
Gene Name
protein kinases;ubiquitin-protein ligase
Gene Aliases
AT5G13530, KEEP ON GOING, T6I14.60, T6I14_60
Chromosome Location
Chr.5: 4345202 - 4354723 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203374.1 NP_001190303.1 12 - 13 2912 74
NM_121356.3 NP_196857.2 12 - 13 3239 74
GenBank mRNA DQ315360.1 - 12 - 13 2823 74

View Full Product Details