Assay Design



Gene Transcripts

Gene Symbol
VAT1
Entrez Gene ID
831490
Gene Name
VALINE-TOLERANT 1
Gene Aliases
AT5G16290, MQK4.1, MQK4_1, VALINE-TOLERANT 1
Chromosome Location
Chr.5: 5333750 - 5337784 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121634.4 NP_197133.1 1 - 2 340 73
NM_180498.3 NP_850829.1 1 - 2 327 73
GenBank mRNA AK226906.1 - 1 - 2 266 73
BT021989.1 - 1 - 2 229 73

View Full Product Details