Assay Design



Gene Transcripts

Gene Symbol
CHR17
Entrez Gene ID
831980
Gene Name
chromatin remodeling factor17
Gene Aliases
AT5G18620, T1A4.2, chromatin remodeling factor17
Chromosome Location
Chr.5: 6195689 - 6202168 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121867.4 NP_568365.2 17 - 18 2462 119
NM_180516.2 NP_850847.1 17 - 18 2460 119

View Full Product Details