Assay Design
Gene Transcripts
- Gene Symbol
- CKX7
- Entrez Gene ID
-
832248
- Gene Name
- cytokinin oxidase 7
- Gene Aliases
-
ARABIDOPSIS THALIANA CYTOKININ OXIDASE 5, AT5G21482, ATCKX5, cytokinin oxidase 7
- Chromosome Location
-
Chr.5: 7226606 - 7230304 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_180532.3 |
NP_850863.1 |
1 - 2 |
955 |
82 |
GenBank mRNA |
AF303981.1 |
- |
1 - 2 |
719 |
82 |
View Full Product Details