Assay Design
Gene Transcripts
- Gene Symbol
- OLI2
- Entrez Gene ID
-
835690
- Gene Name
- S-adenosyl-L-methionine-dependent methyltransferases superfamily protein
- Gene Aliases
-
AT5G55920, OLIGOCELLULA 2
- Chromosome Location
-
Chr.5: 22645448 - 22649517 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_180596.2 |
NP_850927.1 |
1 - 2 |
727 |
70 |
View Full Product Details