Assay Design
Gene Transcripts
- Gene Symbol
- SIR
- Entrez Gene ID
-
830336
- Gene Name
- sulfite reductase
- Gene Aliases
-
AT5G04590, SULFITE REDUCTASE, T1E3.2, sulfite reductase
- Chromosome Location
-
Chr.5: 1319074 - 1323364 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120541.4 |
NP_196079.1 |
6 - 7 |
2050 |
73 |
GenBank mRNA |
AF325027.2 |
- |
6 - 7 |
1764 |
73 |
View Full Product Details