Assay Design



Gene Transcripts

Gene Symbol
CPK1
Entrez Gene ID
830366
Gene Name
calcium dependent protein kinase 1
Gene Aliases
AT5G04870, ATCPK1, CALCIUM DEPENDENT PROTEIN KINASE, calcium dependent protein kinase 1
Chromosome Location
Chr.5: 1416564 - 1420554 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120569.3 NP_196107.1 1 - 2 1591 131
GenBank mRNA AK118706.1 - 1 - 2 1375 131
BT005932.1 - 1 - 2 914 131
L14771.1 - 1 - 2 1063 131

View Full Product Details