Assay Design
Gene Transcripts
- Gene Symbol
- ALA1
- Entrez Gene ID
-
830375
- Gene Name
- aminophospholipid ATPase 1
- Gene Aliases
-
AT5G04930, aminophospholipid ATPase 1
- Chromosome Location
-
Chr.5: 1444638 - 1449879 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120575.3 |
NP_568146.1 |
6 - 7 |
4134 |
100 |
GenBank mRNA |
AF175769.1 |
- |
6 - 7 |
3338 |
100 |
View Full Product Details