Assay Design



Gene Transcripts

Gene Symbol
SDN2
Entrez Gene ID
830437
Gene Name
small RNA degrading nuclease 2
Gene Aliases
AT5G05540, MOP10.8, MOP10_8, small RNA degrading nuclease 2
Chromosome Location
Chr.5: 1636282 - 1638939 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125703.2 NP_001119175.1 4 - 5 506 77
NM_120636.4 NP_196173.1 4 - 5 506 77
GenBank mRNA AK118437.1 - 4 - 5 482 77

View Full Product Details