Assay Design



Gene Transcripts

Gene Symbol
ASA1
Entrez Gene ID
830457
Gene Name
anthranilate synthase alpha subunit 1
Gene Aliases
A-METHYL TRYPTOPHAN RESISTANT 1, AMT1, ANTHRANILATE SYNTHASE ALPHA SUBUNIT, AT5G05730, JASMONATE-INDUCED DEFECTIVE LATERAL ROOT 1, JDL1, MJJ3.14, MJJ3_14, TRP5, TRYPTOPHAN BIOSYNTHESIS 5, WEAK ETHYLENE INSENSITIVE 2, WEI2, anthranilate synthase alpha subunit 1
Chromosome Location
Chr.5: 1719533 - 1722933 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203302.1 NP_001190231.1 7 - 8 1558 76
NM_120655.4 NP_196192.1 8 - 9 1494 76

View Full Product Details