Assay Design
Gene Transcripts
- Gene Symbol
- WOX7
- Entrez Gene ID
-
830462
- Gene Name
- WUSCHEL related homeobox 7
- Gene Aliases
-
AT5G05770, MJJ3.18, MJJ3_18, WOX5A, WUSCHEL related homeobox 5A, WUSCHEL related homeobox 7
- Chromosome Location
-
Chr.5: 1734427 - 1734855 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120659.2 |
NP_196196.1 |
1 - 1 |
384 |
78 |
GenBank mRNA |
AB493739.1 |
- |
1 - 1 |
324 |
78 |
View Full Product Details