Assay Design



Gene Transcripts

Gene Symbol
ATJ6
Entrez Gene ID
830582
Gene Name
J-domain protein 6
Gene Aliases
AT5G06910, J-domain protein 6, MOJ9.8, MOJ9_8
Chromosome Location
Chr.5: 2140400 - 2142679 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120773.4 NP_196308.1 5 - 6 663 84
GenBank mRNA AK228246.1 - 5 - 6 604 84
BT006202.1 - 5 - 6 465 84

View Full Product Details