Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036768.2 | NP_001031845.1 | 8 - 9 | 865 | 67 |
NM_001085070.2 | NP_001078539.1 | 7 - 8 | 819 | 67 | |
NM_001342913.1 | NP_001331392.1 | 8 - 9 | 1260 | 67 | |
NM_120777.3 | NP_196312.1 | 8 - 9 | 1063 | 67 | |
NM_203015.1 | NP_974744.1 | 8 - 9 | 861 | 67 | |
GenBank mRNA | AK117686.1 | - | 8 - 9 | 878 | 67 |
BT006134.1 | - | 6 - 7 | 601 | 67 | |
D10042.1 | - | 8 - 9 | 867 | 67 |