Assay Design
Gene Transcripts
- Gene Symbol
- ST2B
- Entrez Gene ID
-
830591
- Gene Name
- sulfotransferase 2B
- Gene Aliases
-
ARABIDOPSIS THALIANA SULFOTRANSFERASE 2B, AT5G07000, ATST2B, MOJ9.17, MOJ9_17, sulfotransferase 2B
- Chromosome Location
-
Chr.5: 2171367 - 2173015 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_120782.4 |
NP_196317.2 |
1 - 1 |
1531 |
117 |
View Full Product Details