Assay Design



Gene Transcripts

Gene Symbol
scpl35
Entrez Gene ID
830722
Gene Name
serine carboxypeptidase-like 35
Gene Aliases
AT5G08260, F8L15.17, serine carboxypeptidase-like 35
Chromosome Location
Chr.5: 2657054 - 2661437 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120909.4 NP_196443.1 5 - 6 1132 99
GenBank mRNA AF370244.1 - 5 - 6 1018 99
AY062969.1 - 5 - 6 950 99

View Full Product Details