Assay Design



Gene Transcripts

Gene Symbol
HEMC
Entrez Gene ID
830724
Gene Name
hydroxymethylbilane synthase
Gene Aliases
AT5G08280, F8L15.10, F8L15_10, PORPHOBILINOGEN DEAMINASE, RUG1, RUGOSA 1, hydroxymethylbilane synthase
Chromosome Location
Chr.5: 2663499 - 2665826 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120911.4 NP_196445.1 3 - 4 1091 79
GenBank mRNA AF419614.1 - 3 - 4 991 79
AY070431.1 - 3 - 4 960 79
AY123037.1 - 3 - 4 861 79

View Full Product Details