Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001335796.1 | NP_001318266.1 | 5 - 6 | 662 | 97 |
NM_001335797.1 | NP_001324040.1 | 4 - 5 | 823 | 97 | |
NM_127796.3 | NP_565532.1 | 4 - 5 | 648 | 97 | |
GenBank mRNA | AY048261.1 | - | 4 - 5 | 623 | 97 |
AY072625.1 | - | 4 - 5 | 514 | 97 | |
U76846.1 | - | 4 - 5 | 637 | 97 |