Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001335796.1 | NP_001318266.1 | 6 - 7 | 840 | 95 |
NM_001335797.1 | NP_001324040.1 | 5 - 6 | 1001 | 95 | |
NM_127796.3 | NP_565532.1 | 5 - 6 | 826 | 95 | |
GenBank mRNA | AY048261.1 | - | 5 - 6 | 801 | 95 |
AY072625.1 | - | 5 - 6 | 692 | 95 | |
U76846.1 | - | 5 - 6 | 815 | 95 |