Assay Design



Gene Transcripts

Gene Symbol
GRF1
Entrez Gene ID
816815
Gene Name
growth-regulating factor 1
Gene Aliases
AT2G22840, AtGRF1, T20K9.5, T20K9_5, growth-regulating factor 1
Chromosome Location
Chr.2: 9728480 - 9731301 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127849.4 NP_179869.1 3 - 4 1281 111
GenBank mRNA AB493560.1 - 3 - 4 920 111
AY102634.1 - 3 - 4 992 111

View Full Product Details