Assay Design



Gene Transcripts

Gene Symbol
CNGC6
Entrez Gene ID
816931
Gene Name
cyclic nucleotide-gated channel 6
Gene Aliases
AT2G23980, ATCNGC6, CYCLIC NUCLEOTIDE GATED CHANNEL 6, T29E15.18, T29E15_18, cyclic nucleotide-gated channel 6
Chromosome Location
Chr.2: 10201104 - 10205422 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335878.1 NP_001318278.1 5 - 6 1098 78
NM_001335881.1 NP_001323885.1 4 - 5 1445 78
NM_001335882.1 NP_001323879.1 3 - 4 1400 78
NM_001335883.1 NP_001323880.1 2 - 3 1245 78
NM_001335886.1 NP_001323883.1 4 - 5 1053 78
NM_127960.3 NP_565560.1 4 - 5 977 78
GenBank mRNA Y17914.1 - - 877 78

View Full Product Details