Assay Design



Gene Transcripts

Gene Symbol
AO1
Entrez Gene ID
832221
Gene Name
aldehyde oxidase 1
Gene Aliases
AAO1, ALDEHYDE OXIDASE, AOalpha, ARABIDOPSIS THALIANA ALDEHYDE OXIDASE 1, AT-AO1, AT5G20960, ATAO, Arabidopsis thaliana aldehyde oxidase 1, AtAO1, F22D1.130, F22D1_130, aldehyde oxidase 1, aldehyde oxidase alpha
Chromosome Location
Chr.5: 7116455 - 7122774 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122105.3 NP_568407.2 4 - 5 2399 95
NM_180718.2 NP_851049.1 4 - 5 2631 95
GenBank mRNA AB005804.1 - 4 - 5 2348 95

View Full Product Details