Assay Design



Gene Transcripts

Gene Symbol
PHS1
Entrez Gene ID
832437
Gene Name
dual specificity protein phosphatase family protein
Gene Aliases
AT5G23720, MQM1.1, MQM1_1, PROPYZAMIDE-HYPERSENSITIVE 1
Chromosome Location
Chr.5: 7998197 - 8002848 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122277.2 NP_197761.2 1 - 2 68 77
NM_180735.4 NP_851066.2 1 - 2 152 77
GenBank mRNA AB161693.1 - 1 - 2 119 77

View Full Product Details