Assay Design



Gene Transcripts

Gene Symbol
WR3
Entrez Gene ID
835085
Gene Name
nitrate transmembrane transporter
Gene Aliases
AT5G50200, ATNRT3.1, K6A12.6, K6A12_6, NITRATE TRANSPORTER 3.1, NRT3.1, WOUND-RESPONSIVE 3
Chromosome Location
Chr.5: 20436180 - 20437815 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124399.3 NP_199831.1 2 - 3 276 87
NM_180828.2 NP_851159.1 1 - 2 334 87
NM_180829.1 NP_851160.1 2 - 3 285 87
GenBank mRNA AJ311926.1 - 2 - 3 275 87

View Full Product Details