Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161301.1 | NP_001154773.1 | 4 - 5 | 506 | 66 |
NM_001161302.1 | NP_001154774.1 | 4 - 5 | 670 | 66 | |
NM_001344974.1 | NP_001330348.1 | 2 - 3 | 1244 | 66 | |
NM_001344975.1 | NP_001330349.1 | 3 - 4 | 1045 | 66 | |
NM_001344976.1 | NP_001330347.1 | 3 - 4 | 869 | 66 | |
NM_124583.4 | NP_200017.2 | 3 - 4 | 344 | 66 | |
NM_180843.3 | NP_851174.1 | 3 - 4 | 344 | 66 | |
GenBank mRNA | AF370171.1 | - | 3 - 4 | 328 | 66 |
AK319008.1 | - | 4 - 5 | 654 | 66 | |
AY059133.1 | - | 2 - 3 | 225 | 66 |