Assay Design



Gene Transcripts

Gene Symbol
SPDS3
Entrez Gene ID
835392
Gene Name
spermidine synthase 3
Gene Aliases
AT5G53120, ATSPDS3, MFH8.5, MFH8_5, SPERMIDINE SYNTHASE 3, SPMS, spermidine synthase 3
Chromosome Location
Chr.5: 21534473 - 21537816 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085279.1 NP_001078748.1 7 - 8 729 101
NM_001085280.1 NP_001078749.1 6 - 7 634 101
NM_001203598.1 NP_001190527.1 6 - 7 984 101
NM_001345033.1 NP_001332707.1 6 - 7 760 101
NM_124691.3 NP_568785.1 7 - 8 766 101
NM_180847.2 NP_851178.1 6 - 7 826 101
NM_180848.3 NP_851179.1 6 - 7 813 101
GenBank mRNA AY040013.1 - 6 - 7 792 101
AY079367.1 - 5 - 6 548 101
BT000742.1 - 6 - 7 777 101

View Full Product Details