Assay Design



Gene Transcripts

Gene Symbol
SPDS3
Entrez Gene ID
835392
Gene Name
spermidine synthase 3
Gene Aliases
AT5G53120, ATSPDS3, MFH8.5, MFH8_5, SPERMIDINE SYNTHASE 3, SPMS, spermidine synthase 3
Chromosome Location
Chr.5: 21534473 - 21537816 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085279.1 NP_001078748.1 8 - 9 797 118
NM_001085280.1 NP_001078749.1 7 - 8 702 118
NM_001203598.1 NP_001190527.1 7 - 8 1052 118
NM_001345033.1 NP_001332707.1 7 - 8 828 118
NM_124691.3 NP_568785.1 8 - 9 834 118
NM_180847.2 NP_851178.1 7 - 8 894 118
NM_180848.3 NP_851179.1 7 - 8 881 118
GenBank mRNA AY040013.1 - 7 - 8 860 118
AY079367.1 - 6 - 7 616 118
BT000742.1 - 7 - 8 845 118

View Full Product Details