Assay Design
Gene Transcripts
- Gene Symbol
- GIL1
- Entrez Gene ID
-
836013
- Gene Name
- glucose-6-phosphate isomerase, putative (DUF641)
- Gene Aliases
-
AT5G58960, GRAVITROPIC IN THE LIGHT, K19M22.15, K19M22_15
- Chromosome Location
-
Chr.5: 23805717 - 23808478 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_180886.2 |
NP_851217.1 |
1 - 2 |
123 |
74 |
View Full Product Details