Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001125995.2 | NP_001119467.1 | 13 - 14 | 1686 | 115 |
NM_001345427.1 | NP_001332136.1 | 13 - 14 | 1877 | 115 | |
NM_001345428.1 | NP_001332137.1 | 13 - 14 | 1773 | 115 | |
NM_125453.6 | NP_200868.2 | 13 - 14 | 1602 | 115 | |
NM_180902.5 | NP_851233.1 | 13 - 14 | 1686 | 115 | |
GenBank mRNA | AF434673.1 | - | 13 - 14 | 1542 | 115 |