Assay Design
Gene Transcripts
- Gene Symbol
- UBQ1
- Entrez Gene ID
-
824425
- Gene Name
- ubiquitin extension protein 1
- Gene Aliases
-
AT3G52590, EARLY-RESPONSIVE TO DEHYDRATION 16, EMB2167, EMBRYO DEFECTIVE 2167, ERD16, F3C22.8, HAP4, HAPLESS 4, ubiquitin extension protein 1
- Chromosome Location
-
Chr.3: 19505379 - 19506932 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115119.4 |
NP_566969.1 |
4 - 5 |
652 |
96 |
- Gene Symbol
- AT2G36170
- Entrez Gene ID
-
818189
- Gene Name
- 60S ribosomal protein L40-1
- Gene Aliases
-
60S ribosomal protein L40 A, AT2G36170, F9C22.10, F9C22_10, RPL40A, UBQ2, ubiquitin extension protein 2
- Chromosome Location
-
Chr.2: 15172061 - 15173340 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY035148.1 |
- |
4 - 5 |
391 |
96 |
View Full Product Details