Assay Design
Gene Transcripts
- Gene Symbol
- PP2-A15
- Entrez Gene ID
-
824466
- Gene Name
- phloem protein 2-A15
- Gene Aliases
-
AT3G53000, AtPP2-A15, phloem protein 2-A15
- Chromosome Location
-
Chr.3: 19653910 - 19656135 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115160.3 |
NP_190868.1 |
2 - 3 |
796 |
83 |
View Full Product Details