Assay Design



Gene Transcripts

Gene Symbol
GR
Entrez Gene ID
824631
Gene Name
glutathione reductase
Gene Aliases
AT3G54660, ATGR2, EMB2360, glutathione reductase
Chromosome Location
Chr.3: 20229637 - 20233211 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115323.4 NP_191026.1 1 - 2 510 152
GenBank mRNA BT020536.1 - 1 - 2 416 152
BT030335.1 - 1 - 2 399 152
D14049.1 - 1 - 2 417 152

View Full Product Details