Assay Design
Gene Transcripts
- Gene Symbol
- TTN8
- Entrez Gene ID
-
824632
- Gene Name
- Structural maintenance of chromosomes (SMC) family protein
- Gene Aliases
-
AT3G54670, ATSMC1, SMC1, STRUCTURAL MAINTENANCE OF CHROMOSOMES 1, TITAN8
- Chromosome Location
-
Chr.3: 20235506 - 20244046 on Build TAIR10.1
View Full Product Details