Assay Design
Gene Transcripts
- Gene Symbol
- AMP1
- Entrez Gene ID
-
824637
- Gene Name
- Peptidase M28 family protein
- Gene Aliases
-
ALTERED MERISTEM PROGRAM 1, AT3G54720, AtAMP1, CONSTITUTIVE MORPHOGENESIS 2, COP2, HAUPTLING, HPT, MFO1, Multifolia, PRIMORDIA TIMING, PT
- Chromosome Location
-
Chr.3: 20254639 - 20257916 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115329.3 |
NP_567007.1 |
8 - 9 |
1908 |
102 |
GenBank mRNA |
BT046200.1 |
- |
8 - 9 |
1807 |
102 |
View Full Product Details