Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_115349.6 | NP_191052.2 | 3 - 4 | 1773 | 94 |
GenBank mRNA | AF534079.1 | - | 3 - 4 | 1422 | 94 |
AY058870.1 | - | 3 - 4 | 1459 | 94 | |
AY074331.1 | - | 3 - 4 | 1455 | 94 | |
AY096739.1 | - | 3 - 4 | 1247 | 94 |