Assay Design



Gene Transcripts

Gene Symbol
OVA1
Entrez Gene ID
824706
Gene Name
methionyl-tRNA synthetase / methionine-tRNA ligase / MetRS (cpMetRS)
Gene Aliases
AT3G55400, OVULE ABORTION 1
Chromosome Location
Chr.3: 20535631 - 20539138 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084826.2 NP_001078295.1 1 - 2 415 79
NM_115398.3 NP_191100.1 1 - 2 447 79

View Full Product Details