Assay Design



Gene Transcripts

Gene Symbol
TPI
Entrez Gene ID
824710
Gene Name
triosephosphate isomerase
Gene Aliases
AT3G55440, ATCTIMC, CYTOSOLIC ISOFORM TRIOSE PHOSPHATE ISOMERASE, CYTOSOLIC TRIOSE PHOSPHATE ISOMERASE, CYTOTPI, TRIOSEPHOSPHATE ISOMERASE, triosephosphate isomerase
Chromosome Location
Chr.3: 20553403 - 20556560 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115402.4 NP_191104.1 3 - 4 628 88
GenBank mRNA AF375426.1 - 3 - 4 331 88
AY074822.1 - 3 - 4 237 88

View Full Product Details