Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001125374.1 | NP_001118846.1 | 3 - 4 | 346 | 74 |
NM_115419.5 | NP_191120.2 | 3 - 4 | 548 | 74 | |
GenBank mRNA | AY091766.1 | - | 3 - 4 | 346 | 74 |
Y09355.1 | - | - | 316 | 74 |