Assay Design



Gene Transcripts

Gene Symbol
FAR6
Entrez Gene ID
824837
Gene Name
fatty acid reductase 6
Gene Aliases
AT3G56700, AtFAR6, fatty acid reductase 6
Chromosome Location
Chr.3: 20999409 - 21002155 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115529.2 NP_191229.1 4 - 5 730 61
GenBank mRNA DQ056628.1 - 4 - 5 633 61
EU280152.1 - 4 - 5 633 61

View Full Product Details