Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001339805.1 | NP_001326776.1 | 6 - 7 | 1125 | 75 |
NM_115529.2 | NP_191229.1 | 6 - 7 | 1062 | 75 | |
GenBank mRNA | DQ056628.1 | - | 6 - 7 | 965 | 75 |
EU280151.1 | - | 6 - 7 | 1028 | 75 | |
EU280152.1 | - | 6 - 7 | 965 | 75 |