Assay Design



Gene Transcripts

Gene Symbol
AREB3
Entrez Gene ID
824852
Gene Name
ABA-responsive element binding protein 3
Gene Aliases
ABA-responsive element binding protein 3, AT3G56850, DPBF3
Chromosome Location
Chr.3: 21046234 - 21048614 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115544.3 NP_191244.1 2 - 3 1529 105
GenBank mRNA AB017162.1 - 2 - 3 1368 105
AY062448.1 - 2 - 3 1374 105
AY081670.1 - 2 - 3 809 105

View Full Product Details