Assay Design
Gene Transcripts
- Gene Symbol
- LPAT2
- Entrez Gene ID
-
824934
- Gene Name
- lysophosphatidyl acyltransferase 2
- Gene Aliases
-
AT3G57650, lysophosphatidyl acyltransferase 2
- Chromosome Location
-
Chr.3: 21349383 - 21353193 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_115625.4 |
NP_567052.1 |
2 - 3 |
572 |
86 |
GenBank mRNA |
BT026036.1 |
- |
2 - 3 |
204 |
86 |
View Full Product Details