Assay Design



Gene Transcripts

Gene Symbol
NRPA1
Entrez Gene ID
824935
Gene Name
nuclear RNA polymerase A1
Gene Aliases
AT3G57660, nuclear RNA polymerase A1
Chromosome Location
Chr.3: 21353497 - 21363201 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339881.1 NP_001325828.1 14 - 15 2852 90
NM_115626.1 NP_191325.1 14 - 15 2642 90

View Full Product Details