Assay Design



Gene Transcripts

Gene Symbol
UVI4-LIKE
Entrez Gene ID
824955
Gene Name
UV-B-insensitive 4-like protein
Gene Aliases
AT3G57860, GIG1, GIGAS CELL 1, OMISSION OF SECOND DIVISION, OSD1, UV-B-insensitive 4-like
Chromosome Location
Chr.3: 21426285 - 21428340 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115648.5 NP_191345.1 2 - 3 487 60
GenBank mRNA BT004275.1 - 2 - 3 448 60
BT005570.1 - 1 - 2 343 60

View Full Product Details