Assay Design



Gene Transcripts

Gene Symbol
ECT10
Entrez Gene ID
835931
Gene Name
evolutionarily conserved C-terminal region 10
Gene Aliases
AT5G58190, MCK7.6, MCK7_6, evolutionarily conserved C-terminal region 10
Chromosome Location
Chr.5: 23546329 - 23549605 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125204.3 NP_200627.2 6 - 7 1240 92
NM_203225.1 NP_974954.1 6 - 7 1199 92
GenBank mRNA AY072018.1 - 6 - 7 1146 92
BT000833.1 - 6 - 7 1135 92

View Full Product Details