Assay Design



Gene Transcripts

Gene Symbol
BON1
Entrez Gene ID
836311
Gene Name
Calcium-dependent phospholipid-binding Copine family protein
Gene Aliases
AT5G61900, BON, BONZAI 1, COPINE 1, CPN1, K22G18.2, K22G18_2
Chromosome Location
Chr.5: 24855661 - 24859341 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125583.4 NP_568944.1 14 - 15 1505 96
NM_203248.2 NP_974977.1 13 - 14 1540 96
GenBank mRNA AX380787.1 - 13 - 14 1462 96
AY062824.1 - 14 - 15 1496 96
AY085339.1 - 14 - 15 1497 96
BT010555.1 - 13 - 14 1418 96

Gene Symbol
AT5G61910
Entrez Gene ID
836312
Gene Name
DCD (Development and Cell Death) domain protein
Gene Aliases
AT5G61910, K22G18.3, K22G18_3
Chromosome Location
Chr.5: 24855621 - 24864172 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203661.1 NP_001190590.1 20 - 21 3941 96

View Full Product Details