Assay Design



Gene Transcripts

Gene Symbol
HEN4
Entrez Gene ID
836560
Gene Name
RNA-binding KH domain-containing protein
Gene Aliases
AT5G64390, HUA ENHANCER 4, MSJ1.23, MSJ1_23
Chromosome Location
Chr.5: 25743344 - 25748575 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203683.1 NP_001190612.1 3 - 4 1877 140
NM_125835.5 NP_201244.2 3 - 4 1879 140
NM_203261.2 NP_974990.1 3 - 4 1879 140

View Full Product Details