Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_125859.4 | NP_568990.1 | 10 - 11 | 1181 | 67 |
NM_203262.3 | NP_974991.1 | 10 - 11 | 1181 | 67 | |
NM_203263.2 | NP_974992.1 | 10 - 11 | 1096 | 67 | |
GenBank mRNA | AF375435.1 | - | 10 - 11 | 1117 | 67 |
AY074827.1 | - | 10 - 11 | 1048 | 67 |