Assay Design
Gene Transcripts
- Gene Symbol
- PP2-A4
- Entrez Gene ID
-
840289
- Gene Name
- phloem protein 2-A4
- Gene Aliases
-
AT1G33920, ATPP2-A4, T3M13.6, T3M13_6, phloem protein 2-A4
- Chromosome Location
-
Chr.1: 12319905 - 12320870 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103114.1 |
NP_174654.1 |
1 - 2 |
122 |
60 |
View Full Product Details